2,4,4-Trimethyl-2-pentene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
143820-25G 143820-5G
SIAL143820-25GEA 164 EUR
SIAL143820-25G SIAL143820-5G
2,4,4-Trimethyl-2-pentene, Sigma-Aldrich®
2,4,4-Trimethyl-2-pentene
Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 104 °C (1013 hPa)
Melting Pt: –106.3 °C
Density: 0.7223 g/cm³ (20 °C)
MDL Number: MFCD00008902
CAS Number: 107-40-4
EINECS: 203-488-5
UN: 2050
ADR: 3,II