2-Ethyl-1-butene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
E14705-25G E14705-5G
SIALE14705-25GEA 189 EUR
SIALE14705-25G SIALE14705-5G
2-Ethyl-1-butene, Sigma-Aldrich®
2-Ethyl-1-butene
Formula: C₆H₁₂
Boiling Pt: 65 °C (1013 hPa)
Melting Pt: –131.5 °C
Density: 0.688 g/cm³ (24 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009341
CAS Number: 760-21-4
EINECS: 212-078-5
UN: 3295
ADR: 3,II