2-Methyl-2-pentene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,1-Dimethyl-1-butene
M-Clarity™ quality level = MQ100
M67303-25G M67303-5G
SIALM67303-25GEA 721 EUR
SIALM67303-25G SIALM67303-5G
2-Methyl-2-pentene, Sigma-Aldrich®
2-Methyl-2-pentene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 67 °C (1013 hPa)
Melting Pt: –135 °C
Density: 0.6904 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009382
CAS Number: 625-27-4
EINECS: 210-883-6
UN: 3295
ADR: 3,II