3,3-Dimethyl-1-butene, Sigma-Aldrich®
Supplier: Merck
MQ100
119059-250ML 119059-50ML
SIAL119059-250MLEA 200 EUR
SIAL119059-250ML SIAL119059-50ML
3,3-Dimethyl-1-butene, Sigma-Aldrich®
3,3-Dimethyl-1-butene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 41
Melting Pt: –115 °C
Density: 0.6683 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008853
CAS Number: 558-37-2
EINECS: 209-195-9
UN: 1993
ADR: 3 II