trans-Cinnamic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
C80857-250G C80857-5G
SIALC80857-250GEA 102 EUR
SIALC80857-250G SIALC80857-5G
trans-Cinnamic acid, Sigma-Aldrich®
trans-Cinnamic acid
Formula: C₉H₈O₂
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 131…134 °C
Density: 1.25 g/cm³ (4 °C)
MDL Number: MFCD00004369
CAS Number: 140-10-3
EINECS: 205-398-1