3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥97%, Sigma-Aldrich®
Supplier: Merck
Synonyms: Isoferulic acid, 3-(3-Hydroxy-4-methoxyphenyl)acrylic acid
103012-1G 103012-5G
SIAL103012-1GEA 39.2 EUR
SIAL103012-1G SIAL103012-5G
3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥97%, Sigma-Aldrich®
3-Hydroxy-4-methoxycinnamic acid
Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Boiling Pt: 410 °C (1013 hPa)
Melting Pt: 180…182 °C
MDL Number: MFCD00004391
CAS Number: 537-73-5
EINECS: 208-676-0