Sinapic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid
D7927-5G D7927-1G
SIALD7927-5GEA 164 EUR
SIALD7927-5G SIALD7927-1G
Sinapic acid, Sigma-Aldrich®
Sinapic acid
Formula: C₁₁H₁₂O₅
MW: 224.21 g/mol
Boiling Pt: 378
Melting Pt: 200…202 °C
MDL Number: MFCD00004401
CAS Number: 530-59-6
EINECS: 208-487-3