Caffeic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
M-Clarity™ quality level = MQ200
C0625-25G C0625-2G C0625-5G
SIALC0625-25GEA 119 EUR
SIALC0625-25G SIALC0625-2G SIALC0625-5G
Caffeic acid, Sigma-Aldrich®
Caffeic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 234…237 °C
MDL Number: MFCD00004392
CAS Number: 331-39-5
EINECS: 206-361-2
Merck Index: 12,01673