Cinnamyl alcohol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol
M-Clarity™ quality level = MQ100
108197-100G 108197-500G 108197-5G
SIAL108197-100GEA 30.4 EUR
SIAL108197-100G SIAL108197-500G SIAL108197-5G
Cinnamyl alcohol, Sigma-Aldrich®
Cinnamyl alcohol
Formula: C₉H₁₀O
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 31…34 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002921
CAS Number: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325