3-Bromo-1-butyne, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

690643-1G 690643-5G
SIAL690643-1GEA 214 EUR
SIAL690643-1G SIAL690643-5G
3-Bromo-1-butyne, Sigma-Aldrich®
3-Bromo-1-butyne
Formula: C₄H₅Br
MW: 132.99 g/mol
Storage Temperature: Refrigerator
CAS Number: 18668-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR