1,4-Dichloro-2-butyne, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,4-Dichlorobut-2-yne
MQ100
D59607-100G D59607-25G D59607-5G
SIALD59607-100GEA 788 EUR
SIALD59607-100G SIALD59607-25G SIALD59607-5G
1,4-Dichloro-2-butyne, Sigma-Aldrich®
1,4-Dichloro-2-butyne
Formula: C₄H₄Cl₂
MW: 122.98 g/mol
Boiling Pt: 165
Melting Pt: –34.5 °C
Density: 1.2613 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000981
CAS Number: 821-10-3
EINECS: 212-474-8
UN: 3334
ADR: 9