3-Butyne-2-one, Sigma-Aldrich®

Supplier: Merck

Synonyms: But-3-yn-2-one

M-Clarity™ quality level = MQ100

161314-10ML 161314-2.5ML
SIAL161314-10MLEA 557 EUR
SIAL161314-10ML SIAL161314-2.5ML
3-Butyne-2-one, Sigma-Aldrich®
3-Butyne-2-one
Formula: C₄H₄O
MW: 68.08 g/mol
Boiling Pt: 85
Density: 0.886 g/cm³ (15 °C)
MDL Number: MFCD00008775
CAS Number: 1423-60-5
EINECS: 215-834-2
UN: 1992
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR