17-Octadecynoic acid
Supplier: Cayman Chemical
90270-1 90270-10 90270-100 90270-5
CAYM90270-1EA 22 EUR
CAYM90270-1 CAYM90270-10 CAYM90270-100 CAYM90270-5
17-Octadecynoic acid
17-Octadecynoic acid
Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
CAS Number: 34450-18-5