2-Butynoic acid 98%
Supplier: Thermo Fisher Scientific
Danger
308060010 308060050
ACRO308060010EA 43.9 EUR
ACRO308060010 ACRO308060050
2-Butynoic acid 98%
2-Butynoic acid
Formula: C₄H₄O₂
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 74…78 °C
Density: 0,9641 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004363
CAS Number: 590-93-2
EINECS: 209-695-7
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 74 to 78 °C
GC ≥97.5 %