2-Butyne-1-ol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Butyn-1-ol, But-2-yn-1-ol, 1-Hydroxy-2-butyne
M-Clarity™ quality level = MQ200
244341-25G 244341-5G
SIAL244341-25GEA 134 EUR
SIAL244341-25G SIAL244341-5G
2-Butyne-1-ol, Sigma-Aldrich®
2-Butyne-1-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.9483 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002914
CAS Number: 764-01-2
EINECS: 212-113-4
UN: 1987
ADR: 3,III