3-Pentyn-1-ol ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Pent-3-yn-1-ol
L02951.03
L02951.06
L02951.03EA
32
EUR
L02951.03
L02951.06
3-Pentyn-1-ol ≥98%
3-Pentyn-1-ol
Purity: 98%
CAS No.: 10229-10-4
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 54°(129°F)
Refractive Index: 1.4560
Odour: Recognizable
Boiling Point: 154-157°
Beilstein Reference: 1735967
UN No.: 1987
MDL No.: MFCD00002956
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 154…157 °C Density: 0.912 Flash Pt: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002956 CAS Number: 10229-10-4 EINECS: 233-550-7 UN: 1987 ADR: 3,III |