5-Hexyn-1-ol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
302015-1G
302015-25G
302015-5G
SIAL302015-1GEA
44.2
EUR
SIAL302015-1G
SIAL302015-25G
SIAL302015-5G
5-Hexyn-1-ol, Sigma-Aldrich®
5-Hexyn-1-ol
Formula:
C₆H₁₀O MW: 98.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002980 CAS Number: 928-90-5 |