1-Heptyne, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

244414-100ML 244414-25ML
SIAL244414-100MLEA 340 EUR
SIAL244414-100ML SIAL244414-25ML
1-Heptyne, Sigma-Aldrich®
1-Heptyne
Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: –81 °C
Density: 0.7328 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009529
CAS Number: 628-71-7
EINECS: 211-051-5
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR