1-Hexyne, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
244422-100ML 244422-25ML
SIAL244422-100MLEA 134 EUR
SIAL244422-100ML SIAL244422-25ML
1-Hexyne, Sigma-Aldrich®
1-Hexyne
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 71 °C (1013 hPa)
Melting Pt: –132 °C
Density: 0.7147 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009504
CAS Number: 693-02-7
EINECS: 211-736-9
UN: 1993
ADR: 3,II