1-Octyne, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

244465-100G 244465-25G
SIAL244465-100GEA 147 EUR
SIAL244465-100G SIAL244465-25G
1-Octyne, Sigma-Aldrich®
1-Octyne
Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 126
Melting Pt: –1 °C
Density: 0.7461 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009546
CAS Number: 629-05-0
EINECS: 211-069-3
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR