1-Pentyne, Sigma-Aldrich®
Supplier: Merck
MQ100
256560-25G 256560-5G
SIAL256560-25GEA 101 EUR
SIAL256560-25G SIAL256560-5G
1-Pentyne, Sigma-Aldrich®
1-Pentyne
Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 40
Melting Pt: –90 °C
Density: 0.6901 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009469
CAS Number: 627-19-0
EINECS: 210-987-1
UN: 1993
ADR: 3 II