3-Hexyne, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
306894-25G 306894-5G
SIAL306894-25GEA 121 EUR
SIAL306894-25G SIAL306894-5G
3-Hexyne, Sigma-Aldrich®
3-Hexyne
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: –61.5 °C
Density: 0.7231 g/cm³ (20 °C)
Flash Pt: 0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009381
CAS Number: 928-49-4
EINECS: 213-173-4
UN: 1993
ADR: 3,II