Anthracene, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
141062-100G
141062-25G
141062-1G
SIAL141062-100GEA
123
EUR
SIAL141062-100G
SIAL141062-25G
SIAL141062-1G
Anthracene, Sigma-Aldrich®
Anthracene
Formula:
C₁₄H₁₀ MW: 178.23 g/mol Boiling Pt: 340 °C (1013 hPa) Melting Pt: 111…112 °C Density: 1.24 g/cm³ (20 °C) Flash Pt: 121 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001240 CAS Number: 120-12-7 EINECS: 204-371-1 Merck Index: 13,00687 |
|
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...