Anthracene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
A89200-100G A89200-500G
SIALA89200-100GEA 67.8 EUR
SIALA89200-100G SIALA89200-500G
Anthracene, Sigma-Aldrich®
Anthracene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340
Melting Pt: 111…112 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001240
CAS Number: 120-12-7
EINECS: 204-371-1
Merck Index: 13,00687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf973bfe5e591a235e84a414db5e3a617957ab78&vid=39774282d3251d5014028c1ba9d03866f87d7733&prid=web&clid=SAPDAM