9-Phenylanthracene, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
P18004-1G
P18004-5G
SIALP18004-1GEA
66.7
EUR
SIALP18004-1G
SIALP18004-5G
9-Phenylanthracene, Sigma-Aldrich®
9-Phenylanthracene
Formula:
C₂₀H₁₄ MW: 254.33 g/mol Boiling Pt: 417 °C (1013 hPa) Melting Pt: 157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001252 CAS Number: 602-55-1 EINECS: 210-019-8 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...