Quinuclidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Azabicyclo[2.2.2]octane, ABCO

M-Clarity™ quality level = MQ100

197602-10G 197602-1G
SIAL197602-10GEA 842 EUR
SIAL197602-10G SIAL197602-1G
Quinuclidine, Sigma-Aldrich®
Quinuclidine
Formula: C₇H₁₃N
MW: 111.19 g/mol
Boiling Pt: 150
Density: 1.025 g/cm³ (20 °C)
MDL Number: MFCD00006690
CAS Number: 100-76-5
EINECS: 202-887-1
UN: 2811
ADR: 6.1 II
Merck Index: 12,08265

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR