N-Bromosuccinimide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: NBS, 1-Bromopyrrolidine-2,5-dione
M-Clarity™ quality level = MQ200
B81255-100G B81255-1KG B81255-500G B81255-250G B81255-5G
SIALB81255-100GEA 42.2 EUR
SIALB81255-100G SIALB81255-1KG SIALB81255-500G SIALB81255-250G SIALB81255-5G
N-Bromosuccinimide, Sigma-Aldrich®
N-Bromosuccinimide
Formula: C₄H₄BrNO₂
MW: 177.99 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 174…179 °C
Density: 2.098 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005510
CAS Number: 128-08-5
EINECS: 204-877-2
UN: 3261
ADR: 8,III
Merck Index: 13,01423