N-Chlorosuccinimide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1-Chloropyrrolidine-2,5-dione
M-Clarity™ quality level = MQ200
109681-100G
109681-500G
SIAL109681-100GEA
30.8
EUR
SIAL109681-100G
SIAL109681-500G
N-Chlorosuccinimide, Sigma-Aldrich®
N-Chlorosuccinimide
Formula:
C₄H₄ClNO₂ MW: 133.53 g/mol Boiling Pt: 216.5 °C (1013 hPa) Melting Pt: 148…151 °C Density: 1.65 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005511 CAS Number: 128-09-6 EINECS: 204-878-8 UN: 3261 ADR: 8,II Merck Index: 13,02183 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...