2-Pyrrolidone ≥99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Pyrrolidin-2-one
A15332.36 A15332.A3
A15332.36EA 44.1 EUR
A15332.36 A15332.A3
2-Pyrrolidone ≥99%
2-Pyrrolidone
Purity: 99% CAS No.: 616-45-5 Molecular Formula: C4H7NO Formula Weight: 85.11 Flash Point: 110°(230°F) Refractive Index: 1.4875 Melting Point: 23-25° Boiling Point: 245° Merck Reference: 14,8016 MDL No.: MFCD00005270
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 245 °C
Melting Pt: 23…25 °C
Density: 1.12
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005270
CAS Number: 616-45-5
EINECS: 210-483-1
Merck Index: 14,8016