2-Pyrrolidone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Pyrrolidin-2-one
M-Clarity™ quality level = MQ200
83300-1L
83300-250ML
SIAL83300-1LEA
81.1
EUR
SIAL83300-1L
SIAL83300-250ML
2-Pyrrolidone, Sigma-Aldrich®
2-Pyrrolidone
Formula:
C₄H₇NO MW: 85.11 g/mol Boiling Pt: 245 °C (1013 hPa) Melting Pt: 24…26 °C Density: 1.1115 g/cm³ (20 °C) Flash Pt: 129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005270 CAS Number: 616-45-5 EINECS: 210-483-1 Merck Index: 12,08200 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...