Pyrrolidine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Tetrahydropyrrole
MQ100
83240-100ML 83240-25ML 83240-500ML
SIAL83240-100MLEA 51.8 EUR
SIAL83240-100ML SIAL83240-25ML SIAL83240-500ML
Pyrrolidine, Sigma-Aldrich®
Pyrrolidine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 87
Melting Pt: –63 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 3
Storage Temperature: Ambient
MDL Number: MFCD00005249
CAS Number: 123-75-1
EINECS: 204-648-7
UN: 1922
ADR: 3 II
Merck Index: 12,08199