1-Ethyl-2-pyrrolidone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Ethyl-2-pyrrolidinone, N-Ethyl-2-pyrrolidone, 1-Ethylpyrrolidin-2-one
M-Clarity™ quality level = MQ100
146358-500G 146358-100G
SIAL146358-500GEA 357 EUR
SIAL146358-500G SIAL146358-100G
1-Ethyl-2-pyrrolidone, Sigma-Aldrich®
1-Ethyl-2-pyrrolidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 97
Density: 0.9979 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003199
CAS Number: 2687-91-4
EINECS: 220-250-6