1-Methylpiperidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-Methylpiperidine

M-Clarity™ quality level = MQ200

M72609-100ML M72609-500ML
SIALM72609-100MLEA 31 EUR
SIALM72609-100ML SIALM72609-500ML
1-Methylpiperidine, Sigma-Aldrich®
1-Methylpiperidine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 106.5 °C (1013 hPa)
Melting Pt: –69 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 3 °C
Storage Temperature: Freezer
MDL Number: MFCD00006491
CAS Number: 626-67-5
EINECS: 210-959-9
UN: 2399
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR