1-Methylpyrrolidine, Sigma-Aldrich®
Supplier: Merck
Synonyms: N-Methylpyrrolidine
M-Clarity™ quality level = MQ100
69110-100ML 69110-500ML
SIAL69110-100MLEA 51.1 EUR
SIAL69110-100ML SIAL69110-500ML
1-Methylpyrrolidine, Sigma-Aldrich®
1-Methylpyrrolidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 80
Melting Pt: –76 °C
Density: 0.799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003173
CAS Number: 120-94-5
EINECS: 204-438-5
UN: 3286
ADR: 3 II