1-Methylpyrrolidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-Methylpyrrolidine

M-Clarity™ quality level = MQ200

M79204-100ML M79204-500ML M79204-5ML
SIALM79204-100MLEA 51.1 EUR
SIALM79204-100ML SIALM79204-500ML SIALM79204-5ML
1-Methylpyrrolidine, Sigma-Aldrich®
1-Methylpyrrolidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 80 °C (1013 hPa)
Melting Pt: –76 °C
Density: 0.799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003173
CAS Number: 120-94-5
EINECS: 204-438-5
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR