2,2,6,6-Tetramethylpiperidine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
115754-10G 115754-25G 115754-5G
SIAL115754-10GEA 84.1 EUR
SIAL115754-10G SIAL115754-25G SIAL115754-5G
2,2,6,6-Tetramethylpiperidine, Sigma-Aldrich®
2,2,6,6-Tetramethylpiperidine
Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: 30…31 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00005985
CAS Number: 768-66-1
EINECS: 212-199-3