3-Pyrroline, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,5-Dihydro-1H-pyrrole
M-Clarity™ quality level = MQ100
377112-1G 377112-250MG
SIAL377112-1GEA 53.9 EUR
SIAL377112-1G SIAL377112-250MG
3-Pyrroline, Sigma-Aldrich®
3-Pyrroline
Formula: C₄H₇N
MW: 69.11 g/mol
Boiling Pt: 90…91 °C (1013 hPa)
Melting Pt: –71 °C
Density: 0.9097 g/cm³ (20 °C)
MDL Number: MFCD00005213
CAS Number: 109-96-6
EINECS: 203-723-1
Merck Index: 12,08201