Morpholine, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
134236-100ML 134236-2.5L 134236-500ML
SIAL134236-100MLEA 38.4 EUR
SIAL134236-100ML SIAL134236-2.5L SIAL134236-500ML
Morpholine, Sigma-Aldrich®
Morpholine
Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 128.9 °C (1013 hPa)
Melting Pt: –5 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8,I
Merck Index: 12,06362
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fb9ce2adfd770be2ae8d5f63ea6220bef0f781&vid=6b2bfa885631c3f4e0681813373d468dcc171b83&prid=web&clid=SAPDAM