4-Ethylmorpholine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 100-74-3
Molecular Formula: C6H13NO
Formula Weight: 115.18
Flash Point: 27°(80°F)
Refractive Index: 1.4413
Melting Point: -63°
Boiling Point: 138-139°
Beilstein Reference: 102969
UN No.: 2734
MDL No.: MFCD00006177
Formula:
C₆H₁₃NO Boiling Pt: 138…139 °C Melting Pt: –63 °C Density: 0.908 Flash Pt: 27 °C (80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006177 CAS Number: 100-74-3 EINECS: 202-885-0 UN: 2734 ADR: 8,II |