4-Ethylmorpholine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
04499-100ML 04499-25ML
SIAL04499-100MLEA 189 EUR
SIAL04499-100ML SIAL04499-25ML
4-Ethylmorpholine, Sigma-Aldrich®
4-Ethylmorpholine
Formula: C₆H₁₃NO
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00006177
CAS Number: 100-74-3
EINECS: 202-885-0
UN: 2734
ADR: 8,II