4-Ethylmorpholine, Sigma-Aldrich®
Supplier: Merck
Synonyms: N-Ethylmorpholine
MQ100
04500-1L 04500-250ML
SIAL04500-1LEA 88.6 EUR
SIAL04500-1L SIAL04500-250ML
4-Ethylmorpholine, Sigma-Aldrich®
4-Ethylmorpholine
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 138
Melting Pt: –63 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 31
Storage Temperature: Ambient
MDL Number: MFCD00006177
CAS Number: 100-74-3
EINECS: 202-885-0
UN: 2734
ADR: 8 II