2-Methylbenzimidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Methyl-1H-benzo[d]imidazole, 2-Methyl-1H-benzimidazole
M-Clarity™ quality level = MQ100
M29800-100G
M29800-5G
SIALM29800-100GEA
117
EUR
SIALM29800-100G
SIALM29800-5G
2-Methylbenzimidazole, Sigma-Aldrich®
2-Methylbenzimidazole
Formula:
C₈H₈N₂ MW: 132.16 g/mol Boiling Pt: 339 °C (1013 hPa) Melting Pt: 175 °C |
MDL Number:
MFCD00005598 CAS Number: 615-15-6 EINECS: 210-411-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...