Benz[a]anthracene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Benz(alpha)anthracene, BaA, 1,2-Benzanthracene
M-Clarity™ quality level = MQ100
B2209-1G
B2209-500MG
SIALB2209-1GEA
139
EUR
SIALB2209-1G
SIALB2209-500MG
Benz[a]anthracene, Sigma-Aldrich®
Benz[a]anthracene
Formula:
C₁₈H₁₂ MW: 228.29 g/mol Boiling Pt: 437 °C (1013 hPa) Density: 1.245 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003599 CAS Number: 56-55-3 EINECS: 200-280-6 UN: 3077 ADR: 9,III Merck Index: 12,01089 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...