Isatin 98%
Supplier: Thermo Scientific
Synonyms: 2,3-Indolinedione, Indoline-2,3-dione, 1H-Indole-2,3-dione
151491000 151495000
ACRO151491000EA 36.6 EUR
ACRO151491000 ACRO151495000
Isatin 98%
Isatin
Formula: C₈H₅NO₂
MW: 147.13 g/mol
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 201…204 °C
Density: 1,472 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00005718
CAS Number: 91-56-5
EINECS: 202-077-8
Merck Index: 13,05119
Order Now
Specification Test Results
Appearance Orange to red Powder or crystals
Infrared spectrum Conforms
Melting point 199 to 204 °C
Water ≤1.0 % (K.F.)
HPLC ≥97.5 %