Indoline, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,3-Dihydroindol
M-Clarity™ quality level = MQ200
I5605-100G I5605-25G
SIALI5605-100GEA 103 EUR
SIALI5605-100G SIALI5605-25G
Indoline, Sigma-Aldrich®
Indoline
Formula: C₈H₉N
MW: 119.17 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: –21 °C
Density: 1.0676 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005705
CAS Number: 496-15-1
EINECS: 207-816-8