1,4-Naphthoquinone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene
M-Clarity™ quality level = MQ200
70372-1KG 70372-250G 70372-50G
SIAL70372-1KGEA 190 EUR
SIAL70372-1KG SIAL70372-250G SIAL70372-50G
1,4-Naphthoquinone, Sigma-Aldrich®
1,4-Naphthoquinone
Formula: C₁₀H₆O₂
MW: 158.16 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 124 °C
Density: 1.422 g/cm³ (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00001676
CAS Number: 130-15-4
EINECS: 204-977-6
UN: 2811
ADR: 6.1,I
Merck Index: 12,06482