2-Ethoxyphenylboronic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
455520-10G 455520-1G
SIAL455520-10GEA 127 EUR
SIAL455520-10G SIAL455520-1G
2-Ethoxyphenylboronic acid, Sigma-Aldrich®
2-Ethoxyphenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
MDL Number: MFCD00674027
CAS Number: 213211-69-9