3-Methoxyphenylboronic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
(3-Methoxyphenyl)metaboric acid, Dihydroxy(3-methoxyphenyl)borane, 3-Methoxybenzeneboronic acid
M-Clarity™ quality level = MQ100
441686-10G
441686-1G
SIAL441686-10GEA
276
EUR
SIAL441686-10G
SIAL441686-1G
3-Methoxyphenylboronic acid, Sigma-Aldrich®
3-Methoxyphenylboronic acid
Formula:
C₇H₉BO₃ MW: 151.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00161359 CAS Number: 10365-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...