4-Methoxyphenylboronic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
4-Methoxybenzeneboronic acid, Dihydroxy(4-methoxyphenyl)borane, (4-Methoxyphenyl)metaboric acid
M-Clarity™ quality level = MQ200
417599-1G
417599-25G
417599-5G
SIAL417599-1GEA
34.5
EUR
SIAL417599-1G
SIAL417599-25G
SIAL417599-5G
4-Methoxyphenylboronic acid, Sigma-Aldrich®
4-Methoxyphenylboronic acid
Formula:
C₇H₉BO₃ Melting Pt: 204…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039139 CAS Number: 5720-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...