2,4-Dimethoxyphenylboronic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

483486-10G 483486-1G
SIAL483486-10GEA 247 EUR
SIAL483486-10G SIAL483486-1G
2,4-Dimethoxyphenylboronic acid, Sigma-Aldrich®
2,4-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01074590
CAS Number: 133730-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR